Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Acetoxy-2-naphthoic Acid 98.0+%, TCI America™
SDP

Catalog No. A09705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09705G 5 g
A09701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09705G Supplier TCI America Supplier No. A09705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17295-26-0
Molecular Formula C13H10O4
Molecular Weight (g/mol) 230.219
MDL Number MFCD00059556
InChI Key NFTLBCXRDNIJMI-UHFFFAOYSA-N
Synonym 6-acetoxy-2-naphthoic acid, 6-acetyloxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-acetyloxy, 6-acetyloxy naphthalene-2-carboxylic acid, 6-hydroxy-2-naphthoic acid acetate, 6-acetoxy-2-naphthoicacid, acmc-209e5j, 6-acetoxy-2-naphtoic acid, 2-acetoxy-6-naphthoic acid
PubChem CID 87038
IUPAC Name 6-acetyloxynaphthalene-2-carboxylic acid
SMILES CC(=O)OC1=CC2=C(C=C1)C=C(C=C2)C(=O)O

Specifications

Melting Point 228°C
Color White-Yellow
Quantity 5 g
Formula Weight 230.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Acetoxy-2-naphthoic Acid
EINECSNumber (4)-1402
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.