Learn More
Cayman Chemical 6-alkyn NAD sodIum salt 700ug
SDP
Catalog No. 502916802
Encompass_Preferred
A clickable form of NAD+; has been used in conjunction with fluorescent tags in the detection of PARP protein substrates
Catalog No. 50-291-6802 Supplier Cayman Chemical Supplier No. 38485700 UG
A clickable form of NAD+; has been used in conjunction with fluorescent tags in the detection of PARP protein substrates