Learn More
Medchemexpress LLC 6-Alkynyl fucose 100mg | 935658-91-6 | 100 MG
SDP
Catalog No. 5000228164
Encompass_Preferred
6-alkynyl fucose is a bioorthogonal fucose analog used to study protein O-fucosylation and for click-chemistry labeling. It contains a terminal alkyne that participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC), enabling conjugation to azide-containing probes for detection or enrichment of fucosylated glycoconjugates in biochemical and cell biology research.
  • Bioorthogonal fucose analog for O-fucosylation studies.
  • Terminal alkyne enables CuAAC (click chemistry) conjugation.
  • High purity suitable for research applications.
  • Available in multiple package sizes, including 100 MG.
  • Molecular weight 342.30 and formula C15H18O9.
Catalog No. 50-002-28164 Supplier Medchemexpress LLC Supplier No. HY115495100MG
6-alkynyl fucose is a bioorthogonal fucose analog used to study protein O-fucosylation and for click-chemistry labeling. It contains a terminal alkyne that participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC), enabling conjugation to azide-containing probes for detection or enrichment of fucosylated glycoconjugates in biochemical and cell biology research.
  • Bioorthogonal fucose analog for O-fucosylation studies.
  • Terminal alkyne enables CuAAC (click chemistry) conjugation.
  • High purity suitable for research applications.
  • Available in multiple package sizes, including 100 MG.
  • Molecular weight 342.30 and formula C15H18O9.