Learn More
Medchemexpress LLC 6-Alkynyl fucose 10mg | 935658-91-6 | 342.30 | C15H18O9 | 10 MG
SDP
Catalog No. 5000307505
Encompass_Preferred
6-Alkynyl fucose is a bioorthogonal fucose analog bearing an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (click chemistry). It is used as a probe and inhibitor to label, image, and study protein O-fucosylation in biochemical and cellular assays.
  • Contains an alkyne functional group for CuAAC labeling.
  • Useful for imaging and tracking O-fucosylation in cells.
  • Acts as a probe and inhibitor of fucosylation pathways in biological assays.
  • High purity suitable for research applications (98.0%).
  • Available in small-scale quantities for experimental work.
Catalog No. 50-003-07505 Supplier Medchemexpress LLC Supplier No. HY11549510MG
6-Alkynyl fucose is a bioorthogonal fucose analog bearing an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (click chemistry). It is used as a probe and inhibitor to label, image, and study protein O-fucosylation in biochemical and cellular assays.
  • Contains an alkyne functional group for CuAAC labeling.
  • Useful for imaging and tracking O-fucosylation in cells.
  • Acts as a probe and inhibitor of fucosylation pathways in biological assays.
  • High purity suitable for research applications (98.0%).
  • Available in small-scale quantities for experimental work.