Learn More
Medchemexpress LLC 6-Alkynyl fucose 25mg | 935658-91-6 | 342.30 g/mol | C15H18O9 | 25 MG
SDP
Catalog No. 5000228453
Encompass_Preferred
6-Alkynyl fucose is an alkyne-tagged fucose analog used as a chemical probe for studying protein O-fucosylation and for click-chemistry labeling via copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is supplied as a high-purity research compound suitable for biochemical and cell-based assays.
  • Alkyne handle enables CuAAC click labeling and conjugation.
  • Useful probe for O-fucosylation studies and imaging.
  • High purity suitable for biochemical and cell-based assays.
  • Stable as a powder at -20°C for extended storage.
  • Offered in small research quantities for method development and optimization.
Catalog No. 50-002-28453 Supplier Medchemexpress LLC Supplier No. HY11549525MG
6-Alkynyl fucose is an alkyne-tagged fucose analog used as a chemical probe for studying protein O-fucosylation and for click-chemistry labeling via copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is supplied as a high-purity research compound suitable for biochemical and cell-based assays.
  • Alkyne handle enables CuAAC click labeling and conjugation.
  • Useful probe for O-fucosylation studies and imaging.
  • High purity suitable for biochemical and cell-based assays.
  • Stable as a powder at -20°C for extended storage.
  • Offered in small research quantities for method development and optimization.