Learn More
Medchemexpress LLC 6-Alkynyl fucose 50mg | 935658-91-6 | 342.30 g/mol | C15H18O9 | 50 MG
SDP
Catalog No. 5000228165
Encompass_Preferred
6-Alkynyl fucose is a bioorthogonal fucose analog used to study protein O-fucosylation and to label fucosylated glycoconjugates via copper-catalyzed azide-alkyne cycloaddition (CuAAC). It serves as a clickable probe for detection, enrichment, and mechanistic studies of fucosylation and has been used as an inhibitor in Notch signaling research.
  • Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Enables labeling and enrichment of O-fucosylated proteins in biochemical and cell-based assays.
  • High purity (98.0%), suitable for analytical and preparative use.
  • Stable as a powder at -20°C for long-term storage; limited stability in solution.
  • Available in small laboratory pack sizes for convenient experimental use.
Catalog No. 50-002-28165 Supplier Medchemexpress LLC Supplier No. HY11549550MG
6-Alkynyl fucose is a bioorthogonal fucose analog used to study protein O-fucosylation and to label fucosylated glycoconjugates via copper-catalyzed azide-alkyne cycloaddition (CuAAC). It serves as a clickable probe for detection, enrichment, and mechanistic studies of fucosylation and has been used as an inhibitor in Notch signaling research.
  • Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Enables labeling and enrichment of O-fucosylated proteins in biochemical and cell-based assays.
  • High purity (98.0%), suitable for analytical and preparative use.
  • Stable as a powder at -20°C for long-term storage; limited stability in solution.
  • Available in small laboratory pack sizes for convenient experimental use.