Learn More
Medchemexpress LLC 6-Alkynyl fucose 5mg | 935658-91-6 | 342.30 g·mol⁻1 | C15H18O9 | 5 MG
SDP
Catalog No. 5000307654
Encompass_Preferred
6-Alkynyl fucose is a bioorthogonal fucose analog for studying protein O-fucosylation. It features an alkyne functional group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for labeling, detection, and enrichment of fucosylated glycoconjugates.
  • Contains a terminal alkyne compatible with CuAAC click chemistry.
  • Enables metabolic labeling of O-fucosylated proteins.
  • Facilitates enrichment and visualization of fucosylated glycans and proteins.
  • Provided as a high-purity solid suitable for biochemical assays.
  • Packaged in a small quantity for lab-scale experiments.
Catalog No. 50-003-07654 Supplier Medchemexpress LLC Supplier No. HY1154955MG
6-Alkynyl fucose is a bioorthogonal fucose analog for studying protein O-fucosylation. It features an alkyne functional group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for labeling, detection, and enrichment of fucosylated glycoconjugates.
  • Contains a terminal alkyne compatible with CuAAC click chemistry.
  • Enables metabolic labeling of O-fucosylated proteins.
  • Facilitates enrichment and visualization of fucosylated glycans and proteins.
  • Provided as a high-purity solid suitable for biochemical assays.
  • Packaged in a small quantity for lab-scale experiments.