Learn More
Carbosynth LLC. 6-Alkynyl-Fucose (0.5 mg vial), 1ea, CAS# 935658-91-6
SDP
Catalog No. 501685740
Encompass_Preferred
Synonyms: 6-Alk-Fuc; 6-Alkynyl-Fucose Tetraacetate; 1,2,3,4-Tetra-O-Acetyl-6,7-Dideoxy-L-Galacto-Hept-6-Ynopyranose, Application: Fucose Probe for Glycan Imaging / Fucosylation Inhibitor, Storage Temperature: -20°C, References: T.-L. Hsu, S. R. Hanson, K. Kishikawa, S.-K. Wang, M. Sawa, and C.-H. Wong, Proc. Natl. Acad. Sci. U.S.A., 104, 2614 (2007). (Original), Y. Kizuka, M. Nakano, Y. Yamaguchi, K. Nakajima, R. Oka, K. Sato, C.-T. Ren, T.-L. Hsu, C.-H. Wong, and N. Taniguchi, Cell Chem. Biol., 24, 1 (2017). (Pharmacol.)
Catalog No. 50-168-5740 Supplier Carbosynth LLC. Supplier No. CKG23007V0.5MG
Add to Cart
Add to Cart
Synonyms: 6-Alk-Fuc; 6-Alkynyl-Fucose Tetraacetate; 1,2,3,4-Tetra-O-Acetyl-6,7-Dideoxy-L-Galacto-Hept-6-Ynopyranose, Application: Fucose Probe for Glycan Imaging / Fucosylation Inhibitor, Storage Temperature: -20°C, References: T.-L. Hsu, S. R. Hanson, K. Kishikawa, S.-K. Wang, M. Sawa, and C.-H. Wong, Proc. Natl. Acad. Sci. U.S.A., 104, 2614 (2007). (Original), Y. Kizuka, M. Nakano, Y. Yamaguchi, K. Nakajima, R. Oka, K. Sato, C.-T. Ren, T.-L. Hsu, C.-H. Wong, and N. Taniguchi, Cell Chem. Biol., 24, 1 (2017). (Pharmacol.)