Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Alpha-Methylprednisolone Crystalline MP Biomedicals

Catalog No. ICN15167191 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15167191 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15167191 Supplier MP Biomedicals, Inc Supplier No. 0215167191

missing translation for 'validMainframeMsg'

Specifications

CAS 83-43-2
Molecular Formula C22H30O5
Quantity 1 g
MDL Number MFCD00010591
Synonym methylprednisolone, medrol, medrone, urbason, methylprednisolon, 6alpha-methylprednisolone, metilbetasone, medrate, metilprednisolone, dopomedrol
InChI Key VHRSUDSXCMQTMA-PJHHCJLFSA-N
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)CO)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]2(C)C=CC(=O)C=C12
IUPAC Name (1R,3aS,3bS,5S,9aR,9bS,10S,11aS)-1,10-dihydroxy-1-(2-hydroxyacetyl)-5,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Molecular Weight (g/mol) 374.48
PubChem CID 6741
ChEBI CHEBI:6888
Formula Weight 374.5
Physical Form Crystalline
Chemical Name or Material 6-Alpha-Methylprednisolone
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.