Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Amino-1-hexanol | 4048-33-3 | 117.19 | 25 G
SDP

Catalog No. 5000448141
Encompass_Preferred

6-Amino-1-hexanol is a biochemical reagent utilized in life science research as a biological material or organic compound. It also functions as a pharmaceutical intermediate.

  • Used as a biochemical reagent
  • Utilized as a biological material or organic compound
  • Functions as a pharmaceutical intermediate
  • Appears as an off-white to yellow solid
  • Molecular weight of 117.19
  • Purity of 99.78%

Catalog No. 50-004-48141 Supplier Medchemexpress LLC Supplier No. 25GHYW007957
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Amino-1-hexanol is a biochemical reagent utilized in life science research as a biological material or organic compound. It also functions as a pharmaceutical intermediate.

  • Used as a biochemical reagent
  • Utilized as a biological material or organic compound
  • Functions as a pharmaceutical intermediate
  • Appears as an off-white to yellow solid
  • Molecular weight of 117.19
  • Purity of 99.78%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.