Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Amino-1-hexanol

Catalog No. 502886460 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-288-6460 100 g
A5635325G 25 g
A563535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-288-6460 Supplier Sigma Aldrich Supplier No. A56353100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

6-Amino-1-hexanol
TRUSTED_SUSTAINABILITY

Specifications

CAS 4048-33-3
Melting Point 54°C to 58°C (lit.)
Boiling Point 135°C to 140°C (30 mmHg)
Molecular Formula C6H15NO
Linear Formula H2N(CH2)6 OH
MDL Number MFCD00008241
Quantity 100 g
Molecular Weight (g/mol) 117.19
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.