Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-amino-2-[(2S)-pentan-2-yl]oxy-9-(5-piperidin-1-ylpentyl)-7H-purin-8-one | 1207629-49-9 | >98.0% | 390.52 g·mol⁻¹ | C20H34N6O2 | 100 MG
SDP

Catalog No. 5000227074
Encompass_Preferred

GSK2245035 is a selective Toll-Like Receptor 7 (TLR7) agonist used in immune research with preferential type-1 interferon (IFN) stimulating activity. The compound is supplied as an off-white to light-yellow solid for laboratory use and is offered in 100 mg quantities.

  • Selective TLR7 agonist with preferential type-1 interferon stimulation.
  • Molecular weight: 390.52 g·mol⁻¹.
  • Chemical formula: C20H34N6O2.
  • Purity ≥98%.
  • Storage: powder at -20°C for long-term stability.

Catalog No. 50-002-27074 Supplier Medchemexpress LLC Supplier No. HY118250100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK2245035 is a selective Toll-Like Receptor 7 (TLR7) agonist used in immune research with preferential type-1 interferon (IFN) stimulating activity. The compound is supplied as an off-white to light-yellow solid for laboratory use and is offered in 100 mg quantities.

  • Selective TLR7 agonist with preferential type-1 interferon stimulation.
  • Molecular weight: 390.52 g·mol⁻¹.
  • Chemical formula: C20H34N6O2.
  • Purity ≥98%.
  • Storage: powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.