Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-2,4-xylenol 98.0+%, TCI America™
SDP

Catalog No. A096225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A096225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A096225G Supplier TCI America Supplier No. A096225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41458-65-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00007691
InChI Key GISWNAMJAQRJPC-UHFFFAOYSA-N
Synonym 6-Amino-2,4-dimethylphenol, 2-Hydroxy-3,5-dimethylaniline
PubChem CID 100530
IUPAC Name 2-amino-4,6-dimethylphenol
SMILES CC1=CC(C)=C(O)C(N)=C1

Specifications

Melting Point 130°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 6-Amino-2,4-xylenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.