Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-2-methyl-3-nitropyridine, 95%, Thermo Scientific™

Catalog No. AAH6121803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6121803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6121803 Supplier Thermo Scientific Chemicals Supplier No. H6121803

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22280-62-2
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00053582
InChI Key BGMZTBKXOFFTBJ-UHFFFAOYSA-N
Synonym 2-amino-6-methyl-5-nitropyridine, 6-amino-3-nitro-2-picoline, 2-amino-5-nitro-6-methylpyridine, 6-amino-2-methyl-3-nitropyridine, 2-amino-5-nitro-6-methyl pyridine, 2-amino-5-nitro-6-picoline, 6-amino-3-nitro-2-methylpyridine, 6-methyl-5-nitro-2-pyridinamine, 6-methyl-5-nitro-pyridin-2-ylamine, 6-methyl-5-nitro-2-pyridylamine
PubChem CID 247976
IUPAC Name 6-methyl-5-nitropyridin-2-amine
SMILES CC1=C(C=CC(=N1)N)[N+](=O)[O-]

Specifications

Melting Point 185°C to 189°C
Color Yellow
Quantity 1 g
Formula Weight 153.14
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 6-Amino-2-methyl-3-nitropyridine
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.