Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-2-methyl-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. A23261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23261G 1 g
A23265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23261G Supplier TCI America Supplier No. A23261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22280-62-2
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00053582
InChI Key BGMZTBKXOFFTBJ-UHFFFAOYSA-N
Synonym 2-amino-6-methyl-5-nitropyridine, 6-amino-3-nitro-2-picoline, 2-amino-5-nitro-6-methylpyridine, 6-amino-2-methyl-3-nitropyridine, 2-amino-5-nitro-6-methyl pyridine, 2-amino-5-nitro-6-picoline, 6-amino-3-nitro-2-methylpyridine, 6-methyl-5-nitro-2-pyridinamine, 6-methyl-5-nitro-pyridin-2-ylamine, 6-methyl-5-nitro-2-pyridylamine
PubChem CID 247976
IUPAC Name 6-methyl-5-nitropyridin-2-amine
SMILES CC1=C(C=CC(=N1)N)[N+](=O)[O-]

Specifications

Melting Point 191°C
Color Yellow
Quantity 1 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2-methyl-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.