Learn More
Sigma Aldrich 6-Amino-2-(methylsulfanyl)-4(3H)-pyrimidinone
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- This product is a 60wt% solution of 2,2-dimethoxyacetaldehyde (glyoxal dimethyl acetal) in water
- The aldol condensation of 2,2-dimethoxyacetaldehyde with acetoacetic ester in the absence of the catalyst and solvent has been reported
- It participates in the synthesis of isoxazoline vinyl ester pseudopeptides and tetrahydro-α-carboline derivatives of barbituric acid analogs
Specifications
Specifications
| CAS | 1074-41-5 |
| Molecular Formula | C5H7N3OS |
| Linear Formula | C5H7N3OS |
| Quantity | 100 mL |
| MDL Number | MFCD00051034 |
| Molecular Weight (g/mol) | 157.19 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.