Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-3-cyano-2-methylpyridine, 97%

Catalog No. AAH2600003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2600003 1 g
AAH2600006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2600003 Supplier Thermo Scientific Chemicals Supplier No. H2600003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 183428-90-2 | C7H7N3 | 133.154 g/mol

6-Amino-3-cyano-2-methylpyridine is a useful nitrile building block for proteomics research and synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-3-cyano-2-methylpyridine is a useful nitrile building block for proteomics research and synthesis.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183428-90-2
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD05865116
InChI Key OZAIJUZOQOBQDW-UHFFFAOYSA-N
Synonym 6-amino-2-methylnicotinonitrile, 6-amino-3-cyano-2-methylpyridine, 2-amino-5-cyano-6-methylpyridine, 3-pyridinecarbonitrile, 6-amino-2-methyl, 6-amino-2-methyl-nicotinonitrile, pubchem14519, acmc-20ahz0, 6-amino-2-methyl-pyridine-3-carbonitrile, 6-amino-2-methyl nicotinonitrile, 2-methyl-3-cyano-6-amino-pyridine
PubChem CID 2794662
IUPAC Name 6-amino-2-methylpyridine-3-carbonitrile
SMILES CC1=C(C=CC(=N1)N)C#N

Specifications

Melting Point 164°C to 166°C
Quantity 1 g
UN Number UN3439
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 6-Amino-3-cyano-2-methylpyridine
missing translation for 'healthHazard1' H302+H312+H332
missing translation for 'healthHazard2' GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.