Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-3-methyl-1,3-benzoxazol-2(3H)-one, 90%

Catalog No. AC433470050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC433470050 5 g
AC433470010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC433470050 Supplier Thermo Scientific Chemicals Supplier No. 433470050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 99584-10-8 | C8H8N2O2 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99584-10-8
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.16
MDL Number MFCD07801206
InChI Key FPNLXQSOWBNXCN-UHFFFAOYSA-N
Synonym 6-amino-3-methyl-3h-benzooxazol-2-one, 6-amino-3-methyl-1,3-benzoxazol-2 3h-one, 6-amino-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one, 6-amino-3-methyl-3-hydrobenzoxazol-2-one, 6-amino-3-methylbenzoxazol-2-one, 2 3h-benzoxazolone, 6-amino-3-methyl, 6-amino-3-methylbenzo d oxazol-2 3h-one
PubChem CID 6486006
IUPAC Name 6-amino-3-methyl-1,3-benzoxazol-2-one
SMILES CN1C(=O)OC2=CC(N)=CC=C12

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 148°C to 151°C
Color Tan
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Quantity 5 g
Formula Weight 164.17
Percent Purity 90%
Physical Form Powder
Chemical Name or Material 6-Amino-3-methyl-1, 3-benzoxazol-2(3H)-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.