Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Amino-4-ethyl-2H-1,4-benzoxazin-3(4H)-one

Catalog No. OTV000561 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000561 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000561 Supplier Sigma Aldrich Supplier No. OTV000561250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 6-phenylhexanoic acid is an arylalkanoic acid.6-phenylhexanoic acid was employed as a model compound to investigate the effects of chromophore orientation and molecular conformation on surface-enhanced raman scattering based on metal nanostructures
  • It was also employed as a substrate to study the stoichiometry of side chain metabolism or complete mineralization of surrogate napthenic acids under methanogenic conditions

Specifications

CAS 103361-44-0
Molecular Formula C10H12N2O2
Linear Formula C10H12N2O2
Quantity 1 g
MDL Number MFCD09907440
Molecular Weight (g/mol) 192.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.