Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-4-hydroxy-2-naphthalenesulfonic Acid 97.0+%, TCI America™
SDP

Catalog No. A037125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A037125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A037125G Supplier TCI America Supplier No. A037125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 90-51-7
Molecular Formula C10H9NO4S
Quantity 25 g
MDL Number MFCD00003973
Synonym gamma Acid, 2-Amino-8-naphthol-6-sulfonic Acid, 7-Amino-1-naphthol-3-sulfonic Acid
InChI Key HBZVNWNSRNTWPS-UHFFFAOYSA-N
SMILES C1=CC(=CC2=C(C=C(C=C21)S(=O)(=O)O)O)N
IUPAC Name 6-amino-4-hydroxynaphthalene-2-sulfonic acid
Molecular Weight (g/mol) 239.245
PubChem CID 7022
Formula Weight 239.25
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-4-hydroxy-2-naphthalenesulfonic Acid
Show More Show Less
EINECSNumber (4)-1100
RTECSNumber QK1295250
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.