Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-amino-6-deoxy-β-cyclodextrin | 29390-67-8 | MFCD03452964 | 95.0% | 1134.00 | C42H71NO34 | 1 G
SDP

Catalog No. 5000283681
Encompass_Preferred

6-Amino-6-deoxy-β-cyclodextrin is an amino-functionalized derivative of β-cyclodextrin supplied as a solid reagent for research and synthesis. It is commonly used to prepare other cyclodextrin derivatives and as a chiral selector in analytical separations.

  • Amino-functionalized β-cyclodextrin for derivatization and host-guest chemistry.
  • Useful as a chiral selector for capillary electrophoresis separations.
  • Supplied as a solid reagent suitable for laboratory synthesis.
  • Typical purity 95.0% (HPLC).
  • Molecular weight 1,134.00 g/mol.
  • Store at 4°C under nitrogen and protect from moisture.

Catalog No. 50-002-83681 Supplier Medchemexpress LLC Supplier No. HYW1293941G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Amino-6-deoxy-β-cyclodextrin is an amino-functionalized derivative of β-cyclodextrin supplied as a solid reagent for research and synthesis. It is commonly used to prepare other cyclodextrin derivatives and as a chiral selector in analytical separations.

  • Amino-functionalized β-cyclodextrin for derivatization and host-guest chemistry.
  • Useful as a chiral selector for capillary electrophoresis separations.
  • Supplied as a solid reagent suitable for laboratory synthesis.
  • Typical purity 95.0% (HPLC).
  • Molecular weight 1,134.00 g/mol.
  • Store at 4°C under nitrogen and protect from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.