Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-amino-6-deoxy-β-cyclodextrin | 29390-67-8 | MFCD03452964 | 95.0% | 1134.00 g/mol | C42H71NO34 | 100 MG
SDP

Catalog No. 5000282930
Encompass_Preferred

6-Amino-6-deoxy-β-cyclodextrin is an amino-substituted β-cyclodextrin supplied for research use. It functions as a host molecule in supramolecular chemistry and as a building block for conjugation or derivatization where a primary hydroxyl is replaced by an amino group.

  • Provides an amino functional handle for derivatization.
  • Suitable as a host molecule for host-guest and supramolecular studies.
  • Purity 95.0%.
  • Molecular formula C42H71NO34; molecular weight 1134.00 g/mol.
  • Available in small research packs with COA and SDS available.
  • Store under nitrogen at 4°C; in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-82930 Supplier Medchemexpress LLC Supplier No. HYW129394100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Amino-6-deoxy-β-cyclodextrin is an amino-substituted β-cyclodextrin supplied for research use. It functions as a host molecule in supramolecular chemistry and as a building block for conjugation or derivatization where a primary hydroxyl is replaced by an amino group.

  • Provides an amino functional handle for derivatization.
  • Suitable as a host molecule for host-guest and supramolecular studies.
  • Purity 95.0%.
  • Molecular formula C42H71NO34; molecular weight 1134.00 g/mol.
  • Available in small research packs with COA and SDS available.
  • Store under nitrogen at 4°C; in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.