Learn More
Medchemexpress LLC 6-amino-6-deoxy-β-cyclodextrin | 29390-67-8 | MFCD03452964 | 95.0% | 1134.003 g/mol | C42H71NO34 | 500 MG
SDP
Catalog No. 5000283682
Encompass_Preferred
6-Amino-6-deoxy-β-cyclodextrin is a β-cyclodextrin derivative bearing a primary amine at the 6-position, provided as a research-grade reagent for conjugation chemistry, host-guest studies, and formulation research. It is a white to off-white powder with defined purity and molecular weight suitable for use as a building block in supramolecular and drug delivery applications.
  • Provides a primary amine handle for conjugation and functionalization.
  • Suitable for inclusion complex and host-guest chemistry studies.
  • Available in multiple vial sizes for small-scale and preparative work.
  • Supplied with a certificate of analysis and safety data sheet.
  • Stable when stored under nitrogen and away from moisture.
Catalog No. 50-002-83682 Supplier Medchemexpress LLC Supplier No. HYW129394500MG
6-Amino-6-deoxy-β-cyclodextrin is a β-cyclodextrin derivative bearing a primary amine at the 6-position, provided as a research-grade reagent for conjugation chemistry, host-guest studies, and formulation research. It is a white to off-white powder with defined purity and molecular weight suitable for use as a building block in supramolecular and drug delivery applications.
  • Provides a primary amine handle for conjugation and functionalization.
  • Suitable for inclusion complex and host-guest chemistry studies.
  • Available in multiple vial sizes for small-scale and preparative work.
  • Supplied with a certificate of analysis and safety data sheet.
  • Stable when stored under nitrogen and away from moisture.