Learn More
Medchemexpress LLC 6-amino-7,9-dihydro-2-(2-methoxyethoxy)-9-(phenylmethyl)-8H-purin-8-one | 226907-52-4 | MFCD21648470 | 99.4% | 315.33 g/mol | C15H17N5O3 | 5MG
SDP
Catalog No. 5000235277
Encompass_Preferred
SM-360320 (CL-087) is a small-molecule Toll-like receptor 7 (TLR7) agonist (CAS 226907-52-4) with immunomodulatory and antitumor activity. Supplied as a white to off-white powder for research use, it is intended for studies requiring TLR7 activation in vitro and in vivo. Handle according to recommended storage and solvent stability conditions.
Catalog No. 50-002-35277 Supplier Medchemexpress LLC Supplier No. HY1253905MG
SM-360320 (CL-087) is a small-molecule Toll-like receptor 7 (TLR7) agonist (CAS 226907-52-4) with immunomodulatory and antitumor activity. Supplied as a white to off-white powder for research use, it is intended for studies requiring TLR7 activation in vitro and in vivo. Handle according to recommended storage and solvent stability conditions.