Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-7-deazapurine hydrogen sulfate, 97%, Thermo Scientific™

Catalog No. AAH64602MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64602MD 250 mg
AAH6460203 1 g
AAH6460206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64602MD Supplier Thermo Scientific Chemicals Supplier No. H64602MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769951-32-8
Molecular Formula C6H8N4O4S
Molecular Weight (g/mol) 232.214
MDL Number MFCD09787504
InChI Key ZVFLCRGXDSPVKK-UHFFFAOYSA-N
Synonym 4-amino-7h-pyrrolo 2,3-d pyrimidine hydrogen sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulphate, 4-amino-7h-pyrrolo 2,3-d pyrimidine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine; sulfuric acid, 7-deazaadenine; sulfuric acid, 1h-pyrrolo 2,3-d pyrimidin-4-amine sulfate, 7h-pyrrolo 2,3-d pyrimidin-4-amine sulfuric acid, 4-amino-7h-pyrrolo 2,3-d pyrimidinehydrogensulfate, ksc376q6n
PubChem CID 11470294
IUPAC Name 7H-pyrrolo[2,3-d]pyrimidin-4-amine;sulfuric acid
SMILES C1=CNC2=C1C(=NC=N2)N.OS(=O)(=O)O

Specifications

Color Brown
Quantity 250 mg
Formula Weight 232.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Amino-7-deazapurine hydrogen sulfate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.