Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-amino-9-benzyl-2-(2-methoxyethoxy)-7H-purin-8-one | 226907-52-4 | MFCD21337856 | 99.4% | 315.33 g/mol | C15H17N5O3 | 25MG
SDP

Catalog No. 5000235951
Encompass_Preferred

A potent, orally active TLR7 agonist used as an immuno-modulator with reported antitumor activity and adjunct activity in vaccine studies. Supplied as a white to off-white solid, it is used in research to stimulate TLR7-mediated immune responses and to evaluate antiviral and antitumor mechanisms.

  • High purity: 99.4%.
  • Molecular formula: C15H17N5O3.
  • Molecular weight: 315.33 g/mol.
  • Appearance: white to off-white solid.
  • Function: potent, orally active TLR7 agonist and immuno-modulator.
  • Applications: antitumor studies, vaccine adjuvant research, and antiviral assays.
  • Packaging: available in multiple solid pack sizes and DMSO solution formats for experimental use.

Catalog No. 50-002-35951 Supplier Medchemexpress LLC Supplier No. HY12539025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A potent, orally active TLR7 agonist used as an immuno-modulator with reported antitumor activity and adjunct activity in vaccine studies. Supplied as a white to off-white solid, it is used in research to stimulate TLR7-mediated immune responses and to evaluate antiviral and antitumor mechanisms.

  • High purity: 99.4%.
  • Molecular formula: C15H17N5O3.
  • Molecular weight: 315.33 g/mol.
  • Appearance: white to off-white solid.
  • Function: potent, orally active TLR7 agonist and immuno-modulator.
  • Applications: antitumor studies, vaccine adjuvant research, and antiviral assays.
  • Packaging: available in multiple solid pack sizes and DMSO solution formats for experimental use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.