Learn More
Medchemexpress LLC 6′-Amino-D-luciferin | 161055-47-6 | 98.5% | 10 MG
SDP
Catalog No. 5000342174
Encompass_Preferred
6′-Amino-D-luciferin is a derivative of D-luciferin where an amino group replaces the 6-hydroxyl group. It is a natural substrate of luciferase (Luc) and can be used for IVIS imaging in cell/animal experiments. This product is for research use only and not sold to patients.
  • Derivative of D-luciferin
  • Amino group replaces the 6-hydroxyl group
  • Natural substrate of luciferase (Luc)
  • Used for IVIS imaging in cell/animal experiments
Catalog No. 50-003-42174 Supplier Medchemexpress LLC Supplier No. HY12699410MG
6′-Amino-D-luciferin is a derivative of D-luciferin where an amino group replaces the 6-hydroxyl group. It is a natural substrate of luciferase (Luc) and can be used for IVIS imaging in cell/animal experiments. This product is for research use only and not sold to patients.
  • Derivative of D-luciferin
  • Amino group replaces the 6-hydroxyl group
  • Natural substrate of luciferase (Luc)
  • Used for IVIS imaging in cell/animal experiments