Learn More
Medchemexpress LLC 6'-Amino-D-luciferin | 161055-47-6 | 98.5% | 25 MG
SDP
Catalog No. 5000341396
Encompass_Preferred
6'-Amino-D-luciferin is a derivative of D-luciferin with an amino group replacing the 6-hydroxyl group. It serves as a natural substrate of luciferase (Luc) and is utilized for IVIS imaging in various cell and animal experimental settings.
  • Derivative of D-luciferin
  • Features an amino group replacing the 6-hydroxyl group
  • Natural substrate for luciferase (Luc)
  • Applicable for IVIS imaging in cell and animal experiments
  • Red-shifts bioluminescence spectrum of green-yellow emitting luciferase
  • Blue-shifts bioluminescence spectrum of red-emitting luciferase
  • Used to analyze interaction of binding mutant active sites (λmax=588 nm)
  • Capable of detecting apoptotic signals in tumor models
Catalog No. 50-003-41396 Supplier Medchemexpress LLC Supplier No. 25MGHY126994
6'-Amino-D-luciferin is a derivative of D-luciferin with an amino group replacing the 6-hydroxyl group. It serves as a natural substrate of luciferase (Luc) and is utilized for IVIS imaging in various cell and animal experimental settings.
  • Derivative of D-luciferin
  • Features an amino group replacing the 6-hydroxyl group
  • Natural substrate for luciferase (Luc)
  • Applicable for IVIS imaging in cell and animal experiments
  • Red-shifts bioluminescence spectrum of green-yellow emitting luciferase
  • Blue-shifts bioluminescence spectrum of red-emitting luciferase
  • Used to analyze interaction of binding mutant active sites (λmax=588 nm)
  • Capable of detecting apoptotic signals in tumor models