Learn More
Medchemexpress LLC 6′-amino-D-luciferin | 161055-47-6 | MFCD29912852 | ≥95.0% | 279.34 g/mol | C11H9N3O2S2 | 5 MG
SDP
Catalog No. 5000264092
Encompass_Preferred
6′-Amino-D-luciferin is a derivative of D-luciferin with the 6-position hydroxyl replaced by an amino group. It acts as a luciferase substrate for bioluminescence assays and is commonly used for IVIS imaging in cell and animal studies. The compound is supplied as a solid with defined storage recommendations and standard analytical characterization for laboratory use.
  • Natural substrate for luciferase enabling bioluminescent assays.
  • Suitable for IVIS imaging in cell and animal experiments.
  • Solid, light yellow to brown appearance for easy handling.
  • Stable under recommended cold storage conditions for long-term use.
  • Molecular weight 279.34 g/mol and CAS 161055-47-6 for identification.
Catalog No. 50-002-64092 Supplier Medchemexpress LLC Supplier No. HY1269945MG
6′-Amino-D-luciferin is a derivative of D-luciferin with the 6-position hydroxyl replaced by an amino group. It acts as a luciferase substrate for bioluminescence assays and is commonly used for IVIS imaging in cell and animal studies. The compound is supplied as a solid with defined storage recommendations and standard analytical characterization for laboratory use.
  • Natural substrate for luciferase enabling bioluminescent assays.
  • Suitable for IVIS imaging in cell and animal experiments.
  • Solid, light yellow to brown appearance for easy handling.
  • Stable under recommended cold storage conditions for long-term use.
  • Molecular weight 279.34 g/mol and CAS 161055-47-6 for identification.