Learn More
Abcam 6-Amino-D-luciferin, 5MG
Catalog No. 50996108
Encompass_Preferred
MW 279.3 Da. Amino analog of the common D-luciferin substrate. Cell permeable reagent that measures firefly luciferase activity. Ideal for the design of proluminescent bioconjugates (peptidase substrates) of luciferin used in ultrasensitive luminescent assays.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-108 Supplier Abcam Supplier No. AB2743125MG
Add to Cart
Add to Cart
MW 279.3 Da. Amino analog of the common D-luciferin substrate. Cell permeable reagent that measures firefly luciferase activity. Ideal for the design of proluminescent bioconjugates (peptidase substrates) of luciferin used in ultrasensitive luminescent assays.
The product is subject to the following: Abcam Restricted Use Statement