Learn More
Medchemexpress LLC 6-Amino-N-(2-(diethylamino)ethyl)quinoline-2-carboxamide | 1429047-67-5 | 99.7% | C16H22N4O | 100 MG
SDP
Catalog No. 5000358472
Encompass_Preferred
6-Amino-N-(2-(diethylamino)ethyl)quinoline-2-carboxamide is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds. It is supplied as a solid with high purity.
  • High purity: 99.7%
  • Stable as powder at -20°C for 3 years, or 4°C for 2 years
  • Soluble in DMSO at 100 mg/mL
  • Used for the synthesis of active compounds
Catalog No. 50-003-58472 Supplier Medchemexpress LLC Supplier No. HY48173100MG
6-Amino-N-(2-(diethylamino)ethyl)quinoline-2-carboxamide is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds. It is supplied as a solid with high purity.
  • High purity: 99.7%
  • Stable as powder at -20°C for 3 years, or 4°C for 2 years
  • Soluble in DMSO at 100 mg/mL
  • Used for the synthesis of active compounds