Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Amino-N-(2-(diethylamino)ethyl)quinoline-2-carboxamide | 1429047-67-5 | 99.7% | C16H22N4O | 25 MG
SDP

Catalog No. 5000357600
Encompass_Preferred

6-Amino-N-(2-(diethylamino)ethyl)quinoline-2-carboxamide is a drug intermediate used for the synthesis of various active compounds. This product is intended for research use only and has not been fully validated for medical applications.

  • Appears as an off-white to light yellow solid
  • Purity of 99.7%
  • Soluble in DMSO at 100 mg/mL, requiring ultrasonic assistance
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-57600 Supplier Medchemexpress LLC Supplier No. 25MGHY48173
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Amino-N-(2-(diethylamino)ethyl)quinoline-2-carboxamide is a drug intermediate used for the synthesis of various active compounds. This product is intended for research use only and has not been fully validated for medical applications.

  • Appears as an off-white to light yellow solid
  • Purity of 99.7%
  • Soluble in DMSO at 100 mg/mL, requiring ultrasonic assistance
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.