Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Amino-N-(2-(diethylamino)ethyl)quinoline-2-carboxamide | 1429047-67-5 | 99.7% | C16H22N4O | 250 MG
SDP

Catalog No. 5000358023
Encompass_Preferred

6-Amino-N-(2-(diethylamino)ethyl)quinoline-2-carboxamide is a drug intermediate used for the synthesis of various active compounds. It is an off-white to light yellow solid intended for research use only.

  • Purity: 99.7%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Molecular weight: 286.38
  • Chemical formula: C16H22N4O
  • CAS number: 1429047-67-5

Catalog No. 50-003-58023 Supplier Medchemexpress LLC Supplier No. 250MGHY48173
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Amino-N-(2-(diethylamino)ethyl)quinoline-2-carboxamide is a drug intermediate used for the synthesis of various active compounds. It is an off-white to light yellow solid intended for research use only.

  • Purity: 99.7%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Molecular weight: 286.38
  • Chemical formula: C16H22N4O
  • CAS number: 1429047-67-5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.