Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminobenzo[b]thiophene-1,1-dioxide, Thermo Scientific™

Catalog No. MWP00044DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MWP00044DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MWP00044DA Supplier Thermo Scientific Chemicals Supplier No. MWP00044DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20503-40-6
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD00173800
InChI Key KRUCRVZSHWOMHC-UHFFFAOYSA-N
Synonym 6-aminobenzo b thiophene 1,1-dioxide, 6-amino-1h-1lambda-6-benzo b thiophene-1,1-dione, 6-amino-1,1-dioxobenzo b thiophene, benzo b thiophen-6-amine,1,1-dioxide, 6-amino-1lambda6-benzothiophene-1,1-dione, 6-amino-1??-benzothiophene-1,1-dione, 6-amino-1, pubchem14478, maybridge1_005192, 6-aminobenzo b thiole-1,1-dione
PubChem CID 2779855
IUPAC Name 1,1-dioxo-1-benzothiophen-6-amine
SMILES C1=CC(=CC2=C1C=CS2(=O)=O)N

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 181.21
Percent Purity Tech.
Grade Technical
Chemical Name or Material 6-Aminobenzo[b]thiophene 1, 1-dioxide
Hazard Category Warning
Hazard Statement GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.