Learn More
Cayman Chemical 6-Aminocaproic Acid, Purity: ≥95%, CAS: 60-32-2, 50 g
SDP
Catalog No. 502994430
Encompass_Preferred
An inhibitor of the plasmin activating enzymes streptokinase, fibrinokinase, and urokinase (IC50s = ~8 μM for all in an enzyme assay); selective for plasmin activating enzymes over trypsin (IC50 = >500 μM); inhibits in vitro plasmin activation and fibrinolysis in a dose-dependent manner; reduces ex vivo fibrinolysis in dog plasma following i.p. administration at doses ranging from 20-100 mg/kg
Catalog No. 50-299-4430 Supplier Cayman Chemical Supplier No. 2372450 G
An inhibitor of the plasmin activating enzymes streptokinase, fibrinokinase, and urokinase (IC50s = ~8 μM for all in an enzyme assay); selective for plasmin activating enzymes over trypsin (IC50 = >500 μM); inhibits in vitro plasmin activation and fibrinolysis in a dose-dependent manner; reduces ex vivo fibrinolysis in dog plasma following i.p. administration at doses ranging from 20-100 mg/kg