Learn More
Apexbio Technology LLC (6-)ε-Aminocaproic acid 60-32-2 50mg
SDP
Catalog No. 502240520
Encompass_Preferred
6-epsilon-Aminocaproic acid is a small-molecule inhibitor targeting plasminogen activators It is designed to competitively inhibit the conversion of plasminogen to plasmin thereby regulating the fibrinolytic pathway and hemostatic processes 6-epsilon-Aminocaproic acid exerts its biological activity primarily through antifibrinolytic inhibition by blocking plasmin-mediated fibrin degradation Based on these pharmacological properties 6-epsilon-Aminocaproic acid holds research potential in coagulation studies thrombosis bleeding disorders and the investigation of pathological fibrinolysis mechanisms
This item is not returnable. View return policy
Catalog No. 50-224-0520 Supplier Apexbio Technology LLC Supplier No. B207650
6-epsilon-Aminocaproic acid is a small-molecule inhibitor targeting plasminogen activators It is designed to competitively inhibit the conversion of plasminogen to plasmin thereby regulating the fibrinolytic pathway and hemostatic processes 6-epsilon-Aminocaproic acid exerts its biological activity primarily through antifibrinolytic inhibition by blocking plasmin-mediated fibrin degradation Based on these pharmacological properties 6-epsilon-Aminocaproic acid holds research potential in coagulation studies thrombosis bleeding disorders and the investigation of pathological fibrinolysis mechanisms