Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Aminocaproic acid | 60-32-2 | 98.0% | 100 G
SDP

Catalog No. 5000405482
Encompass_Preferred

6-Aminocaproic acid (EACA) is a monoamino carboxylic acid that acts as a potent and orally active inhibitor of plasmin and plasminogen. It functions as an antifibrinolytic agent, preventing clot lysis by competitively binding to lysine residues on plasminogen, which in turn inhibits plasmin formation and reduces fibrinolysis. This product is suitable for research related to bleeding disorders.

  • Potent and orally active inhibitor of plasmin and plasminogen.
  • Antifibrinolytic agent that prevents clot lysis.
  • Useful for research of bleeding disorders.
  • Can be used as a hydrophobic linker to improve near-infrared fluorescence imaging and photothermal cancer therapy.
  • Molecular weight of 131.17.
  • Solid, white to off-white appearance.
  • Formula is C6H13NO2.

Catalog No. 50-004-05482 Supplier Medchemexpress LLC Supplier No. HYB0236100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Aminocaproic acid (EACA) is a monoamino carboxylic acid that acts as a potent and orally active inhibitor of plasmin and plasminogen. It functions as an antifibrinolytic agent, preventing clot lysis by competitively binding to lysine residues on plasminogen, which in turn inhibits plasmin formation and reduces fibrinolysis. This product is suitable for research related to bleeding disorders.

  • Potent and orally active inhibitor of plasmin and plasminogen.
  • Antifibrinolytic agent that prevents clot lysis.
  • Useful for research of bleeding disorders.
  • Can be used as a hydrophobic linker to improve near-infrared fluorescence imaging and photothermal cancer therapy.
  • Molecular weight of 131.17.
  • Solid, white to off-white appearance.
  • Formula is C6H13NO2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.