Learn More
Medchemexpress LLC 6-Aminocaproic acid | 60-32-2 | 99.9% | 1 ML
SDP
Catalog No. 5000405517
Encompass_Preferred
6-Aminocaproic acid (EACA), a monoamino carboxylic acid, is a potent and orally active inhibitor of plasmin and plasminogen. It is a potent antifibrinolytic agent that prevents clot lysis through the competitive binding of lysine residues on plasminogen, inhibiting plasmin formation and reducing fibrinolysis. It can be used for the research of bleeding disorders.
  • Potent and orally active inhibitor of plasmin and plasminogen
  • Prevents clot lysis through competitive binding of lysine residues on plasminogen
  • Inhibits plasmin formation and reduces fibrinolysis
  • Useful for research of bleeding disorders
  • Shipping at room temperature in continental US
  • Store at room temperature for 3 years
  • When in solvent, store at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-004-05517 Supplier Medchemexpress LLC Supplier No. HYB023610MM/1ML
6-Aminocaproic acid (EACA), a monoamino carboxylic acid, is a potent and orally active inhibitor of plasmin and plasminogen. It is a potent antifibrinolytic agent that prevents clot lysis through the competitive binding of lysine residues on plasminogen, inhibiting plasmin formation and reducing fibrinolysis. It can be used for the research of bleeding disorders.
  • Potent and orally active inhibitor of plasmin and plasminogen
  • Prevents clot lysis through competitive binding of lysine residues on plasminogen
  • Inhibits plasmin formation and reduces fibrinolysis
  • Useful for research of bleeding disorders
  • Shipping at room temperature in continental US
  • Store at room temperature for 3 years
  • When in solvent, store at -80°C for 2 years or -20°C for 1 year