Learn More
Medchemexpress LLC 6-Aminocaproic acid | 60-32-2 | 99.9% | 100 MG
SDP
Catalog No. 5000407218
Encompass_Preferred
6-Aminocaproic acid (EACA), a monoamino carboxylic acid, is a potent and orally active inhibitor of plasmin and plasminogen. It is also a potent antifibrinolytic agent that prevents clot lysis by competitively binding to lysine residues on plasminogen, thereby inhibiting plasmin formation and reducing fibrinolysis. It can be used for research on bleeding disorders.
  • Potent and orally active inhibitor of plasmin and plasminogen.
  • Antifibrinolytic agent.
  • Prevents clot lysis by inhibiting plasmin formation.
  • Can be used for research of bleeding disorders.
Catalog No. 50-004-07218 Supplier Medchemexpress LLC Supplier No. HYB0236100MG
6-Aminocaproic acid (EACA), a monoamino carboxylic acid, is a potent and orally active inhibitor of plasmin and plasminogen. It is also a potent antifibrinolytic agent that prevents clot lysis by competitively binding to lysine residues on plasminogen, thereby inhibiting plasmin formation and reducing fibrinolysis. It can be used for research on bleeding disorders.
  • Potent and orally active inhibitor of plasmin and plasminogen.
  • Antifibrinolytic agent.
  • Prevents clot lysis by inhibiting plasmin formation.
  • Can be used for research of bleeding disorders.