Learn More
Medchemexpress LLC 6-Aminocaproic acid | 60-32-2 | MFCD00008238 | 99.86% | 500 MG
SDP
Catalog No. 5000405609
Encompass_Preferred
6-Aminocaproic acid (EACA) is a monoamino carboxylic acid that acts as a potent and orally active inhibitor of plasmin and plasminogen. It functions as an antifibrinolytic agent by competitively binding to lysine residues on plasminogen, thereby inhibiting plasmin formation and reducing fibrinolysis. This compound is used in research for bleeding disorders.
  • Potent and orally active inhibitor of plasmin and plasminogen.
  • Functions as an antifibrinolytic agent.
  • Used in research for bleeding disorders.
  • Inhibits fibrinolysis in Asian elephant plasma.
  • Can be used as a hydrophobic linker.
  • Stable under recommended storage conditions.
  • Rapidly absorbed and eliminated in vivo.
Catalog No. 50-004-05609 Supplier Medchemexpress LLC Supplier No. HYB0236500MG
6-Aminocaproic acid (EACA) is a monoamino carboxylic acid that acts as a potent and orally active inhibitor of plasmin and plasminogen. It functions as an antifibrinolytic agent by competitively binding to lysine residues on plasminogen, thereby inhibiting plasmin formation and reducing fibrinolysis. This compound is used in research for bleeding disorders.
  • Potent and orally active inhibitor of plasmin and plasminogen.
  • Functions as an antifibrinolytic agent.
  • Used in research for bleeding disorders.
  • Inhibits fibrinolysis in Asian elephant plasma.
  • Can be used as a hydrophobic linker.
  • Stable under recommended storage conditions.
  • Rapidly absorbed and eliminated in vivo.