Learn More
Medchemexpress LLC 6-[(aminocarbonyl)(2,6-difluorophenyl)amino]-2-(2,4-difluorophenyl)-3-pyridinecarboxamide | 745833-23-2 | MFCD11616590 | 99.7% | 404.32 g/mol | C19H12F4N4O2 | 5 MG
SDP
Catalog No. 5000243608
Encompass_Preferred
VX-702 is a selective small-molecule inhibitor of p38α mitogen-activated protein kinase (MAPK) used in biochemical and cellular research to probe MAPK signaling and inflammatory pathways. It exhibits approximately 14-fold greater potency for p38α versus p38β and is provided as a high-purity research reagent.
  • Selective inhibition of p38α with ~14-fold selectivity over p38β.
  • High purity (≈99.7%), suitable for analytical and cellular studies.
  • Solid form available in milligram quantities for research use.
  • Compatible with common laboratory solvents (e.g., DMSO) for assay preparation.
  • Recommended storage protocols support long-term stability of powder and solutions.
Catalog No. 50-002-43608 Supplier Medchemexpress LLC Supplier No. HY104015MG
VX-702 is a selective small-molecule inhibitor of p38α mitogen-activated protein kinase (MAPK) used in biochemical and cellular research to probe MAPK signaling and inflammatory pathways. It exhibits approximately 14-fold greater potency for p38α versus p38β and is provided as a high-purity research reagent.
  • Selective inhibition of p38α with ~14-fold selectivity over p38β.
  • High purity (≈99.7%), suitable for analytical and cellular studies.
  • Solid form available in milligram quantities for research use.
  • Compatible with common laboratory solvents (e.g., DMSO) for assay preparation.
  • Recommended storage protocols support long-term stability of powder and solutions.