Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminochrysene 97%, Thermo Scientific™

Catalog No. AC103442500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC103442500 250 mg
AC103440010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103442500 Supplier Thermo Scientific Chemicals Supplier No. 103442500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2642-98-0
Molecular Formula C18H13N
Molecular Weight (g/mol) 243.309
InChI Key KIVUHCNVDWYUNP-UHFFFAOYSA-N
Synonym 6-aminochrysene, 6-chrysenamine, chrysenex, chrysonex, 6-amc, chrysen-6-ylamine, unii-i56l81bl2l, ccris 755, chrysene-6-ylamine, 6-chrysenylamine
PubChem CID 17534
IUPAC Name chrysen-6-amine
SMILES C1=CC=C2C(=C1)C=CC3=C2C=C(C4=CC=CC=C34)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 206°C to 211°C
Color Orange to Brown
Infrared Spectrum Authentic
Quantity 250 mg
Beilstein 12, 1346
Merck Index 15, 2258
Formula Weight 243.31
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminochrysene, 97%
Show More Show Less
EINECSNumber 220-149-7
RTECSNumber GC0500000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.