Learn More
AdipoGen 6-Aminofluorescein
SDP
Supplier:  AdipoGen CDXA0019G010
Encompass_Preferred
Chemical. CAS 51649-83-3. Formula C20H13NO5. MW 347.32. Synthetic. Glycosaminoglycuronans react with 6-aminofluorescein to yield fluorescent derivatives.
Catalog No. 50-256-1866