Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminoindole 98.0+%, TCI America™
SDP

Catalog No. A20671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A20671G Supplier TCI America Supplier No. A20671G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 5318-27-4
Melting Point 72°C
Boiling Point 166°C
Molecular Formula C8H8N2
Quantity 1 g
MDL Number MFCD02093959
Synonym 6-aminoindole, 6-amino-1h-indole, 6-amino indole, 1h-indol-6-ylamine, indole-6-ylamine, 1h-indol-6-amin, 6-indolamine, indol-6-amine, pubchem7311
InChI Key MIMYTSWNVBMNRH-UHFFFAOYSA-N
SMILES NC1=CC=C2C=CNC2=C1
IUPAC Name 1H-indol-6-amine
Molecular Weight (g/mol) 132.17
PubChem CID 256096
Formula Weight 132.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminoindole
Show More Show Less
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.