Learn More
Selleck Chemical LLC 6-Aminonicotinamide 25mg
SDP
Supplier:  Selleck Chemical LLC S978325MG
Encompass_Preferred
6-Aminonicotinamide (6AN) is an antimetabolite used to inhibit theNADPH-producing pentose phosphate pathway (PPP) in many cellular systems, making them more susceptible to oxidative stress. 6-Aminonicotinamide is a competitive inhibitor ofNADP+-dependent enzyme glucose-6-phosphate dehydrogenase (G6PD) with Ki of 0.46 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3507