Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-aminonicotinic acid | 3167-49-5 | MFCD00006326 | 99.3% | 138.13 g/mol | C6H6N2O2 | 100 G
SDP

Catalog No. 5000300522
Encompass_Preferred

6-Aminonicotinic acid is a pyridine carboxylic acid used as a research reagent in biochemical and pharmacological studies. It functions as a GABAA receptor agonist (reported Ki ≈ 4.4 nM) and has been described with activities including protein synthesis inhibition and vitamin B3 agonism. The compound is supplied as a solid for laboratory use.

  • Molecular formula: C6H6N2O2.
  • Molecular weight: 138.13 g/mol.
  • CAS number: 3167-49-5.
  • Purity: 99.26%.
  • Appearance: white to light yellow solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Application: for receptor pharmacology and biochemical research.

Catalog No. 50-003-00522 Supplier Medchemexpress LLC Supplier No. HYW015793100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

6-Aminonicotinic acid is a pyridine carboxylic acid used as a research reagent in biochemical and pharmacological studies. It functions as a GABAA receptor agonist (reported Ki ≈ 4.4 nM) and has been described with activities including protein synthesis inhibition and vitamin B3 agonism. The compound is supplied as a solid for laboratory use.

  • Molecular formula: C6H6N2O2.
  • Molecular weight: 138.13 g/mol.
  • CAS number: 3167-49-5.
  • Purity: 99.26%.
  • Appearance: white to light yellow solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Application: for receptor pharmacology and biochemical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.