Learn More
Medchemexpress LLC HY-W013549 500mg Medchemexpress, 6-Aminopenicillanic acid CAS:551-16-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW013549500MG
Encompass_Preferred
Medchemexpress, HY-W013549 500mg 6-Aminopenicillanic acid CAS:551-16-6 6-Aminopenicillanic acid (6-APA) is an important precursor for the synthesis of -lactam antibiotics. 6-Aminopenicillanic acid is the main product of Penicillin G (PenG) hydrolyzed by penicillin acylase (PA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3349