Learn More
AdipoGen 6-Aminophenanthridine (25 mg)
SDP
Supplier:  AdipoGen AGMRC0029M025
Encompass_Preferred
Chemical. CAS: 832-68-8. Formula: C13H10N2. MW: 194.2. Antiprion agent. Ribosome-borne protein folding activity (RPFA) inhibitor. Binds to the ribosomal RNA and inhibits specifically the protein folding activity of the ribosome. Inhibitor of protein aggregation.
Catalog No. 50-205-4250