Learn More
Cayman Chemical 6-AmInophenthrIdIn 10mg
SDP
Catalog No. 502933709
Encompass_Preferred
An antiprion agent; inhibits prion formation in yeast- and mammalian-based screening assays; inhibits protein folding activity of the ribosome at 300 µM; directly competes with protein substrates for the ribosome active site and decreases the yield of refolded protein without affecting the rate of refolding; prevents progressive wing position defects in a Drosophila model of OPMD at 300-400 µM in larvae and 1-3 mM in adults
Catalog No. 50-293-3709 Supplier Cayman Chemical Supplier No. 2642010 MG
An antiprion agent; inhibits prion formation in yeast- and mammalian-based screening assays; inhibits protein folding activity of the ribosome at 300 µM; directly competes with protein substrates for the ribosome active site and decreases the yield of refolded protein without affecting the rate of refolding; prevents progressive wing position defects in a Drosophila model of OPMD at 300-400 µM in larvae and 1-3 mM in adults