Learn More
Medchemexpress LLC 6-aminopyrazolopyrimidine derivative | 1381930-17-1 | 99.4% | 533.62 | C28H35N7O4 | 100MG
SDP
Catalog No. 5000239374
Encompass_Preferred
TBK1/IKKε-IN-4 is a 6-aminopyrazolopyrimidine-derived small-molecule inhibitor selective for TBK1 and IKKε, intended for biochemical and cell-based research into innate immunity and inflammatory signaling. It shows nanomolar potency and is supplied as a solid for laboratory use.
  • Potent TBK1 inhibition, IC50 13 nM.
  • Potent IKKε inhibition, IC50 59 nM.
  • High chemical purity, 99.42%.
  • Solid, white to light yellow appearance.
  • Stable when stored sealed, away from moisture and light; recommended in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in multiple package sizes, including a 100 mg pack.
Catalog No. 50-002-39374 Supplier Medchemexpress LLC Supplier No. HY124652100MG
TBK1/IKKε-IN-4 is a 6-aminopyrazolopyrimidine-derived small-molecule inhibitor selective for TBK1 and IKKε, intended for biochemical and cell-based research into innate immunity and inflammatory signaling. It shows nanomolar potency and is supplied as a solid for laboratory use.
  • Potent TBK1 inhibition, IC50 13 nM.
  • Potent IKKε inhibition, IC50 59 nM.
  • High chemical purity, 99.42%.
  • Solid, white to light yellow appearance.
  • Stable when stored sealed, away from moisture and light; recommended in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in multiple package sizes, including a 100 mg pack.